Anti-DPYSL5 Rabbit Polyclonal Antibody
PRSI15-253
- Antibody type:Primary
- Antigen name:Dihydropyrimidinase like 5
- Antigen symbol:DPYSL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:56896
- Antigen synonyms:CRMP3-associated molecule|CRMP-5|CRMP5|dihydropyrimidinase-related protein 5|collapsin response mediator protein-5|UNC33-like phosphoprotein 6|DRP-5|CRAM|ULIP-6|Ulip6
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 445-564 of human DPYSL5 (NP_064519.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: DPYSL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat