Anti-ABCB10 Rabbit Polyclonal Antibody
Catalog # PRSI15-204
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:ATP-binding cassette, sub-family B (MDR/TAP), member 10
- Antigen symbol:ABCB10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:23456
- Antigen synonyms:MTABC2|M-ABC2|ABCB10|ABC-me|sub-family B|EST20237|ATP-binding cassette|sub-family B (MDR/TAP)|member 10
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 230-300 of human ABCB10 (NP_036221.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ABCB10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat