Anti-RBBP7 Rabbit Polyclonal Antibody
Catalog # PRSI14-998
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Histone-binding protein RBBP7
- Antigen symbol:RBBP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- ChIP:Yes
- Form:Liquid
- Gene ID:5931
- Antigen synonyms:Retinoblastoma-binding protein 7|RBBP7|Histone acetyltransferase type B subunit 2|Retinoblastoma-binding protein p46|RBBP-7|Nucleosome-remodeling factor subunit RBAP46|RBAP46|Histone-binding protein RBBP7
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human RBBP7 (NP_002884.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RBBP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat