Anti-PRKAA1 Rabbit Polyclonal Antibody
PRSI14-346
- Antibody type:Primary
- Antigen name:protein kinase, AMP-activated, alpha 1 catalytic subunit
- Antigen symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:5562
- Antigen synonyms:AMPK|MGC33776|MGC57364|PRKAA1|AMPKa1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human AMPK alpha 1 (NP_006242.5)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat