Anti-IFNGR2 Rabbit Polyclonal Antibody
Catalog # PRSI62-577
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Interferon gamma receptor 2
- Antigen symbol:IFNGR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Epitope:308-337 aa
- Form:Liquid
- Gene ID:3460
- Modification:None
- Molecular weight:38 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This IFNGR2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 308-337 amino acids from the C-terminal region of human IFNGR2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: IFNGR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 308-337
Host: Rabbit
Isotype: Ig
Reactivity: Human