Anti-RB1 Rabbit Polyclonal Antibody
PRSI62-205
- Antibody type:Primary
- Antigen name:Retinoblastoma-associated protein
- Antigen symbol:RB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Epitope:586-615 aa
- Form:Liquid
- Gene ID:5925
- Modification:None
- Molecular weight:106 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,35 mg/ml
- Shipping temperature:4 °C
- Immunogen:This RB1 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 586-615 amino acids from human RB1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000. For IF starting dilution is: 1:10~50. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: RB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 586-615
Host: Rabbit
Isotype: Ig
Reactivity: Human