Anti-FBXO34 Rabbit Polyclonal Antibody
PRSI62-010
- Antibody type:Primary
- Antigen name:F-box only protein 34
- Antigen symbol:FBXO34
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Hamster,Human
- Western blot:Yes
- Epitope:600-629 aa
- Form:Liquid
- Gene ID:55030
- Modification:None
- Molecular weight:79 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,257 mg/ml
- Shipping temperature:4 °C
- Immunogen:This FBXO34 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 600-629 amino acids from the C-terminal region of human FBXO34.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: FBXO34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 600-629
Host: Rabbit
Isotype: Ig
Reactivity: Human, Hamster