Anti-ELP6 Rabbit Polyclonal Antibody
PRSI61-814
- Antibody type:Primary
- Antigen name:Elongator complex protein 6
- Antigen symbol:ELP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:223-252 aa
- Form:Liquid
- Gene ID:54859
- Modification:None
- Molecular weight:30 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,48 mg/ml
- Shipping temperature:4 °C
- Immunogen:This C3orf75 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 223-252 amino acids from the C-terminal region of human C3orf75.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: ELP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 223-252
Host: Rabbit
Isotype: Ig
Reactivity: Human