Anti-XRCC6 Rabbit Polyclonal Antibody
PRSI61-786
- Antibody type:Primary
- Antigen name:X-ray repair cross-complementing protein 6
- Antigen symbol:XRCC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Epitope:521-548 aa
- Form:Liquid
- Gene ID:2547
- Modification:None
- Molecular weight:70 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This XRCC6 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 521-548 amino acids from the C-terminal region of human XRCC6.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50. For IF starting dilution is: 1:10~50.
Type: Primary
Antigen: XRCC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 521-548
Host: Rabbit
Isotype: Ig
Reactivity: Human