Anti-TOX3 Rabbit Polyclonal Antibody
Catalog # PRSI61-675
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:TOX high mobility group box family member 3
- Antigen symbol:TOX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Epitope:214-242 aa
- Form:Liquid
- Gene ID:27324
- Modification:None
- Molecular weight:63 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,48 mg/ml
- Shipping temperature:4 °C
- Immunogen:This TOX3 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 214-242 amino acids from the central region of human TOX3.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: TOX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 214-242
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse