Order Entry
Ireland
ContactUsLinkComponent
Eye Lysate (14 Days Old)
Eye Lysate (14 Days Old)
Catalog # PRSI1420-14
Supplier:  ProSci Inc.
New Product
CAS Number:  
Eye Lysate (14 Days Old)
Catalog # PRSI1420-14
Supplier:  ProSci Inc.
New Product
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    0,1 mg
  • Description:
    Normal; Source: Mouse; Tissue: Eye
  • Storage conditions:
    Store at 2...8 °C for continuous use. For extended storage, freeze working aliquots at −70 °C.
  • Storage buffer:
    Eye lysate (14 Days Old) is supplied in SDS sample buffer containing 5% β-mercaptoethanol
  • Concentration:
    2 mg/ml
  • Tested applications:
    WB

Specifications

About this item

Eye tissue lysate (14 Days Old) was prepared by homogenisation in modified RIPA buffer (150 mM sodium chloride, 50 mM Tris-HCl, pH 7,4, 1 mM ethylenediaminetetraacetic acid, 1 mM phenylmethylsulfonyl fluoride, 1% Triton X-100, 1% sodium deoxycholic acid, 0,1% sodium dodecylsulfate, 5 μg/ml of aprotinin, 5 μg/ml of leupeptin.

Tissue and cell debris was removed by centrifugation. Protein concentration was determined with Bio-Rad protein assay. The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6,8, 12,5% glycerol, 1% sodium dodecylsulfate, 0,01% bromophenol blue) containing 5% β-mercaptoethanol.