Anti-ZNF10 Rabbit Polyclonal Antibody
PRSI59-243
New Product
- Antibody type:Primary
- Antigen name:Zinc finger protein 10
- Antigen symbol:ZNF10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:118 - 148 aa
- Form:Liquid
- Gene ID:7556
- Modification:None
- Molecular weight:66 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This ZNF10 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 118 - 148 amino acids from the N-terminal region of human ZNF10.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ZNF10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 118-148
Host: Rabbit
Isotype: Ig
Reactivity: Human