Anti-ATF5 Rabbit Polyclonal Antibody
Catalog # PRSI58-150
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Cyclic AMP-dependent transcription factor ATF-5
- Antigen symbol:ATF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:188 - 216 aa
- Form:Liquid
- Gene ID:22809
- Modification:None
- Molecular weight:31 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ATF5 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 188-216 amino acids from the C-terminal region of human ATF5
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ATF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 188-216
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse