Order Entry
Ireland
ContactUsLinkComponent
Anti-XPNPEP2 Rabbit Polyclonal Antibody
Anti-XPNPEP2 Rabbit Polyclonal Antibody
Catalog # PRSI13-536
Supplier:  ProSci Inc.
Anti-XPNPEP2 Rabbit Polyclonal Antibody
Catalog # PRSI13-536
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    X-prolyl aminopeptidase 2
  • Antigen symbol:
    XPNPEP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    7512
  • Antigen synonyms:
    AEACEI|APP2|aminoacylproline aminopeptidase|membrane-bound aminopeptidase P|xaa-Pro aminopeptidase 2|membrane-bound APP|mAmP|membrane-bound|X-prolyl aminopeptidase (aminopeptidase P) 2|X-Pro aminopeptidase 2|membrane-bound AmP
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 22-240 of human XPNPEP2 (NP_003390.4)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: XPNPEP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat