Anti-APEX2 Rabbit Polyclonal Antibody
Catalog # PRSI63-726
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:DNA-(apurinic or apyrimidinic site) lyase 2
- Antigen symbol:APEX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:143-171 aa
- Form:Liquid
- Gene ID:27301
- Modification:None
- Molecular weight:57 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This APEX2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 143-171 amino acids from the Central region of human APEX2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IF starting dilution is: 1:25. For IHC-P starting dilution is: 1:10~50. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: APEX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 143-171
Host: Rabbit
Isotype: Ig
Reactivity: Human