Anti-KLRC1 Rabbit Polyclonal Antibody
Catalog # PRSI63-556
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:NKG2-A/NKG2-B type II integral membrane protein
- Antigen symbol:KLRC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:180-206 aa
- Form:Liquid
- Gene ID:3821
- Modification:None
- Molecular weight:26 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,4 mg/ml
- Shipping temperature:4 °C
- Immunogen:This KLRC1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 180-206 amino acids from the C-terminal region of human KLRC1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:200 µl
Specifications
About this item
For IF starting dilution is: 1:25. For FACS starting dilution is: 1:25. For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: KLRC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 180-206
Host: Rabbit
Isotype: Ig
Reactivity: Human