Anti-PDGFRB Rabbit Polyclonal Antibody
Catalog # PRSI63-121
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Platelet-derived growth factor receptor beta
- Antigen symbol:PDGFRB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:40-72 aa
- Form:Liquid
- Gene ID:5159
- Modification:None
- Molecular weight:124 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This PDGFRB antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 40-72 amino acids from the N-terminal region of human PDGFRB.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
Specifications
About this item
For IHC-P starting dilution is: 1:10~50. For IF starting dilution is: 1:10~50. For FACS starting dilution is: 1:10~50. For WB starting dilution is: 1:1000.
Type: Primary
Antigen: PDGFRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 40-72
Host: Rabbit
Isotype: Ig
Reactivity: Human