Anti-FGFR1 Rabbit Polyclonal Antibody
PRSI63-079
- Antibody type:Primary
- Antigen name:Fibroblast growth factor receptor 1
- Antigen symbol:FGFR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:19-48 aa
- Form:Liquid
- Gene ID:2260
- Modification:None
- Molecular weight:92 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This FGFR1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 19~48 amino acids from the N-terminal region of human FGFR1.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
For FACS starting dilution is: 1:10~50. For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For IF starting dilution is: 1:50~100.
Type: Primary
Antigen: FGFR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 19-48
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse