Anti-PRKAA2 Rabbit Polyclonal Antibody
Catalog # PRSI62-889
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:5'-AMP-activated protein kinase catalytic subunit alpha-2
- Antigen symbol:PRKAA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:453-483 aa
- Form:Liquid
- Gene ID:5563
- Modification:None
- Molecular weight:62 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This AMPK alpha2 (PRKAA2) antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 453-483 amino acids from the C-terminal region of human AMPK alpha2 (PRKAA2).
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: PRKAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 453-483
Host: Rabbit
Isotype: Ig
Reactivity: Human