Anti-ADCY1 Rabbit Polyclonal Antibody
Catalog # PRSI23-902
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:adenylate cyclase 1
- Antigen symbol:ADCY1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:107
- Antigen synonyms:AC1|adenylyl cyclase 1|adenylate cyclase type 1|adenylate cyclase type I|3'|ATP pyrophosphate-lyase 1|DFNB44|adenyl cyclase|Ca(2+)/calmodulin-activated adenylyl cyclase|5'-cyclic AMP synthetase|adenylate cyclase 1 (brain)
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 470-610 of human ADCY1 (NP_066939.1)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ADCY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse