Order Entry
Ireland
ContactUsLinkComponent
 
Anti-APITD1 Rabbit Polyclonal Antibody
Catalog # PRSI23-493
Supplier:  ProSci Inc.
New Product
Anti-APITD1 Rabbit Polyclonal Antibody
Catalog # PRSI23-493
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Centromere protein S
  • Antigen symbol:
    APITD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    378708
  • Antigen synonyms:
    APITD1|CENPS|CENP-S|FAAP16|Apoptosis-inducing TAF9-like domain-containing protein 1|FANCM-interacting histone fold protein 1|Fanconi anemia-associated polypeptide of 16 kDa|Centromere protein S|MHF1
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-138 of human APITD1 (NP_954988.1)
  • Purification:
    Affinity purification.
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: APITD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human