Anti-PRDM4 Recombinant Antibody [Clone: RAB-C367]
Catalog # PRSI24-848
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:PR/SET domain 4
- Antigen symbol:PRDM4
- Clonality:Recombinant
- Clone:RAB-C367
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:11108
- Antigen synonyms:PRDM4-A003-1436|EC:2.1.1.-|PR domain-containing protein 4|anti-PRDM4-RAB-C367
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM4 protein under non-denaturing conditions
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: PR/SET domain 4
Clonality: Recombinant
Clone: RAB-C367
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human