Anti-Ley Recombinant Antibody [Clone: H18A]
Catalog # PRSI24-409
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:H18A
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Lewis Y
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Lewis
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: Ley
Clonality: Recombinant
Clone: H18A
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human