Vanquish Rain Trousers
: Portwest
A complimentary trouser style to tie into the PWR rainwear collection. The vanquish trouser uses the highly waterproof and breathable Portwest extreme fabric with taped seams to ensure full protection.
- Abrasion-resistant oxford polyester
- 2 side zip pockets for secure storage
- Elastic waist with toggle for a secure and comfortable fit
- Side zip leg opening for easy access
- Double-layer reinforced knee protection for extra strength and durability
- Taped seams to provide additional protection
- Fully elasticated waistband for ultimate wearer comfort
Durable, breathable, windproof and water-resistant fabric. The extremely water resistant eco-conscious fabric finish beads water away from the fabric surface. A zipped side leg allows for easy access on and off while wearing boots. A fully elasticated, adjustable waistband and articulated legs ensure an excellent fit.
Recommended for wet weather wear. Black finish.
: EN 343 Class 3:3 X WP 10,000 mm, MVP 15,000 g/M²/24 hr. CE certified.
- Catalog No:
- PRTWS556BKRS
- PRTWS556BKRXXL
- PRTWS556BKRXL
- PRTWS556BKRM
- PRTWS556BKRL
- Garment application:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Gender:
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Fabric:
- 100% polyester 600D Oxford, PU coated
- 100% polyester 600D Oxford, PU coated
- 100% polyester 600D Oxford, PU coated
- 100% polyester 600D Oxford, PU coated
- 100% polyester 600D Oxford, PU coated
- Weight:
- 0,57 g/m²
- 0,668 g/m²
- 0,646 g/m²
- 0,592 g/m²
- 0,592 g/m²
Frequently Bought Together