Trypsin
Catalog # PROOFIP13
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- CAS No.:9002-07-7
- Source:
- Species:
- Environmentally Preferable:
- Storage conditions:Refrigerator
- MDL No.:MFCD00082094
- Enzyme name:Trypsin