Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl red
Methyl red
Methyl red
Methyl red
VWRUVW3434-2
:
VWR Collection
:
Methyl red
VWRUVW3434-2
:
VWR Collection
:
Pack type:
Glass bottle
Pk:
500 mL
Seq:
2725
:
:
12,06199
:
178…182 °C
:
1,31 g/cm³ (20 °C)
:
269.3 g/mol
:
Ambient
:
207-776-1
:
493-52-7
:
C₁₅H₁₅N₃O₂
:
MFCD00002425