Order Entry
Ireland
ContactUsLinkComponent
Herbicide reference standard, Trifluralin 100 µg/ml in methanol
Herbicide reference standard, Trifluralin 100 µg/ml in methanol
Catalog # ACCUP-197S
Supplier:  AccuStandard
CAS Number:  
Herbicide reference standard, Trifluralin 100 µg/ml in methanol
Catalog # ACCUP-197S
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pk:
    1 mL
  • CAS No.:
    1582-09-8
  • Description:
    Trifluralin 100 µg/ml in methanol
  • Formula:
    C₁₃H₁₆F₃N₃O₄
  • Description:
    Trifluralin

Specifications

About this item