Order Entry
Ireland
ContactUsLinkComponent
 
BAPTA-AM ≥90% (by TLC)
Catalog # ENZOBMLCA4110025
Supplier:  Enzo Life Sciences
CAS Number:  
BAPTA-AM ≥90% (by TLC)
Catalog # ENZOBMLCA4110025
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :764.69 g/mol
CAS Number:126150-97-8
Formula:C₃₄H₄₀N₂O₁₈
MDL Number:MFCD00036696