MPLA (from E.coli) ≥99.9% serotype R515 (Re), TLRgrade™ ready-to-use
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
:
Storage Temperature:Refrigerator
Specification Test Results
- Concentration:1 mg/ml
- Formulation:Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
- Purity:≥99.9% (regarding protein or DNA contaminants, KDO not detectable)
Specification Test Results
Frequently Bought Together