MPLA 1mg/ml (from Salmonella minnesota) ≥99.9%, TLRgrade™
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
:
Storage Temperature:Refrigerator
Specification Test Results
- Concentration:1 mg/ml
- Formulation:Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
- Purity:≥99.9% (regarding protein or DNA contaminants, KDO not detectable)
Specification Test Results
Frequently Bought Together