MTEP ≥98% (by NMR)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:329205-68-7
:
Formula:C₁₁H₈N₂S
:200.3 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity (NMR):≥98%
- Identity:Identity determined by 1H-NMR
- Appearance:Pale yellow oil
Specification Test Results
Frequently Bought Together