

Procaspase-3-activator (PAC-1) ≥98% (by HPLC), white to off-white solid
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:315183-21-2
:
Formula:C₂₃H₂₈N₄O₂
MDL Number:MFCD03302441
:392.49 g/mol
UN:3077
ADR:9,III
:GHS07
:GHS09
Storage Temperature:Freezer
Specification Test Results
- Purity (HPLC (UV)):≥98%
Specification Test Results
Frequently Bought Together