Order Entry
Ireland
ContactUsLinkComponent
Ristocetin A sulphate ≥95% (by HPLC)
Ristocetin A sulphate ≥95% (by HPLC)
Catalog # ENZOALX380237M010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Ristocetin A sulphate ≥95% (by HPLC)
Catalog # ENZOALX380237M010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :2067.9 · 98.0 g/mol
Storage Temperature:Freezer
CAS Number:11140-99-1
Formula:C₉₅H₁₁₀N₈O₄₄·H₂SO₄