R-sulphoraphane ≥98% (by UPLC)
Catalog # ENZOALX350230M010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:177.2876 g/mol
CAS Number:142825-10-3
Formula:C₆H₁₁NOS₂
MDL Number:MFCD09039283