Order Entry
Ireland
ContactUsLinkComponent
HA-100 dihydrochloride ≥98% (by HPLC)
HA-100 dihydrochloride ≥98% (by HPLC)
Catalog # ENZOALX270069M005
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
HA-100 dihydrochloride ≥98% (by HPLC)
Catalog # ENZOALX270069M005
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210
  • Environmentally Preferable:

Specifications

About this item

Synonyms: HA-100, 5-(Piperazin-1-ylsulphonyl)isoquinoline
 :
 :277.3421 g/mol
CAS Number:84468-24-6
Formula:C₁₃H₁₅N₃O₂S
MDL Number:MFCD00209852