Order Entry
Ireland
ContactUsLinkComponent
 
SL327 ≥99% (by HPLC)
Catalog # ENZOBMLEI3650010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
SL327 ≥99% (by HPLC)
Catalog # ENZOBMLEI3650010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :335.4 g/mol
Storage Temperature:Freezer
CAS Number:305350-87-2
Formula:C₁₆H₁₂F₃N₃S
MDL Number:MFCD06411432