Nullscript ≥97% (by TLC)
:300816-11-9
:
:C₁₆H₁₄N₂O₄
:MFCD00405081
:298.30 g/mol
Frequently Bought Together
Sampling scoops, LaboPlast®
: BURKLE
Accessories and consumables for KingFisher™ Flex
: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
VWR®, Conical Sample Containers, with Snap-On Lids
: VWR Collection
VWR®, Ice Pans, with or without Lid
: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Petri dishes
: Greiner Bio-One
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
: VWR Collection
Cell culture flasks with standard or filter caps, CELLSTAR®
: Greiner Bio-One
Well caps for U96 DeepWell™ plates, Nunc™
: Thermo Fisher Scientific
Sample Spoons, Disposable, SteriWare®
: Sampling Systems
Inoculating loops, stainless steel
: KOEHLER TECHNISCHE PRODUKTEN