AM-251 ≥98% (by HPLC)
Catalog # ENZOBMLCR1080010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Purity:≥98% (HPLC)
- Identity:Determined by ¹H-NMR and MS
Specification Test Results
About this item
:
:555.2 g/mol
Storage Temperature:Ambient
CAS Number:183232-66-8
Formula:C₂₂H₂₁Cl₂IN₄O
MDL Number:MFCD01861181