Order Entry
Ireland
ContactUsLinkComponent
 
AM-404 ≥98% (by TLC)
Catalog # ENZOBMLCR1060050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
AM-404 ≥98% (by TLC)
Catalog # ENZOBMLCR1060050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :395.587 g/mol
CAS Number:198022-70-7
Formula:C₂₆H₃₇NO₂
MDL Number:MFCD03095721