Gö 6983 ≥95% (by HPLC)
Catalog # ENZOBMLEI2960500
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:0,5 mg
- Seq:2035
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-(1-(3-(Dimethylamino)propyl)-5-methoxy-1H-indol-3-yl)-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione, 3-[1-[3-(dimethylamino)propyl]-5-methoxy-1H-indol-3-yl]-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione, Go 6983
:
:442.519 g/mol
Storage Temperature:Freezer
CAS Number:133053-19-7
Formula:C₂₆H₂₆N₄O₃
MDL Number:MFCD04040031