Order Entry
Ireland
ContactUsLinkComponent
Fura 2-AM ≥95% (by HPLC), Ultrapure
Fura 2-AM ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52006
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Fura 2-AM ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52006
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1001.857 g/mol
Storage Temperature:Freezer
CAS Number:108964-32-5
Formula:C₄₄H₄₇N₃O₂₄
MDL Number:MFCD00036976