Order Entry
Ireland
ContactUsLinkComponent
Modular Universal Actuator
Modular Universal Actuator
Catalog # VICIUMHA
Supplier:  VICI
Modular Universal Actuator
Catalog # VICIUMHA
Supplier:  VICI
Specifications
  • Description:
    Modular universal actuators, UMH high speed with RS232 interface
  • Pk:
    1
Specifications
About this item
The universal actuator allows instrument manufacturers to use a single motor and control software to operate virtually any Valco or Cheminert rotary valve. This modular version gives even more flexibility in space utilisation. An added benefit is that the more sensitive electronics module can be located away from any potential liquid leaks from the valve or fittings.
  • Modular for more flexibility in instrument design
  • Replacement for the microelectric actuator
  • One electric actuator works with two position valves and selectors
  • Simplified, universal communication protocol
  • Variety of interfaces, various valve torque requirements
As with the standard universal actuator, the modular is available in three versions: high speed, medium speed/medium torque, and high torque.
This actuator is a replacement for the microelectric multiposition actuator; the motor is a identical stepper motor/gearbox assembly, and a manual remote control, interconnecting cables. And while the modular universal actuator perfectly mimics the microelectric's functionality, it packs even more features into the same small package.
These actuator provides universal 24 volt DC power supply.
Ordering information: Mounting hardware must be ordered separately.