Nitric acid 0.5 mol/l (0.5 N)
Catalog # 5143.9010
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:10 L
- Seq:5214
- Environmentally Preferable:
Specifications
About this item
Synonyms: Nitric acid solution
:
:63.01 g/mol
Boiling Pt:87
EINECS:231-714-2
UN:2031
:GHS07
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349
Melting Pt:-42
Density:1.51
ADR:8 II
Merck Index:13,06610