Order Entry
Ireland
ContactUsLinkComponent
Mouse SERPINA3C ELISA Kit
Mouse SERPINA3C ELISA Kit
Catalog # AVIVOKWB00278
CAS Number:  
Mouse SERPINA3C ELISA Kit
Catalog # AVIVOKWB00278
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    HRP
  • Host:
    Mouse
  • Target protein:
  • Storage conditions:
  • Sample type:
    Serum, plasma, tissue homogenates, cell cultures, other biological fluids
  • Detection method:
    Colorimetric, OD 450 nm
  • Assay Principle:
    Quantitative
  • Specificity:
  • Sensitivity:
    46.9 pg/ml

Specifications

About this item

Aviva Systems Biology Serpina3c ELISA Kit (Mouse) (OKWB00278) is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody specific for Serpina3c has been pre-coated onto a 96-well plate (12 x 8 Well Strips). Standards or test samples are added to the wells, incubated and removed. A biotinylated detector antibody specific for Serpina3c is added, incubated and followed by washing. Avidin-Peroxidase Conjugate is then added, incubated and unbound conjugate is washed away. An enzymatic reaction is produced through the addition of TMB substrate which is catalyzed by HRP generating a blue color product that changes to yellow after adding acidic stop solution. The density of yellow coloration read by absorbance at 450 nm is quantitatively proportional to the amount of sample Serpina3c captured in the well.

Mean Intra-Assay Precision <10%
Mean Inter-Assay Precision <12%