0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standard, Cholesterol 10 µg/ml in methanol
Organic reference standard, Cholesterol 10 µg/ml in methanol
Organic reference standard, Cholesterol 10 µg/ml in methanol
PROONE9237
:
LGC Standards PROMOCHEM
:
Organic reference standard, Cholesterol 10 µg/ml in methanol
PROONE9237
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
10 mL
CAS No.:
57-88-5
Description:
Cholesterol 10 µg/ml in methanol
Formula:
C₂₇H₄₆O
Description:
Cholesterol