Order Entry
Ireland
ContactUsLinkComponent
Pharmaceutical Standards, N-Nitroso Rasagiline
Pharmaceutical Standards, N-Nitroso Rasagiline
Catalog # 1A04030
Supplier:  USP
CAS Number:  
Pharmaceutical Standards, N-Nitroso Rasagiline
Catalog # 1A04030
Supplier:  USP
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Pharmaceutical analytical impurity
  • CAS No.:
    2470278-90-9
  • Description:
    N-Nitroso rasagiline solution (1 ml (1 mg/ml)) ((R)-N-(2,3-Dihydro-1H-inden-1-yl)-N-(prop-2-yn-1-yl)nitrous amide)
  • Storage conditions:
    Keep cool. Store in a well-ventilated place. Keep container tightly closed. Store locked up.
  • Hazard class description:
    Highly flammable liquid and vapor. Toxic if swallowed, contact with skin, if inhaled. Suspected of causing cancer. Causes damage to organs.
  • UN No.:
    1230
  • GHS signal:
    Danger
  • Formula:
    C₁₂H₁₂N₂O
  • Colour:
    Colorless
  • Type:
    Ampule
  • Physical form:
    Liquid

Specifications

About this item