Order Entry
Ireland
ContactUsLinkComponent
Stainless Steel Pipe Fitting Hexagon Nipple
Stainless Steel Pipe Fitting Hexagon Nipple
Catalog # RADS499-2947
Supplier:  RADIONICS
Stainless Steel Pipe Fitting Hexagon Nipple
Catalog # RADS499-2947
Supplier:  RADIONICS

Specifications

  • Connection Type:
    Male
  • Thread:
    9,52 mm, R (taper)
  • Description:
    Stainless steel hexagon nipple 9,52 mm R(T) Male × 9,52 mm R(T) Male, 32 mm
  • Length:
    32,004 mm
  • Pk:
    1

Specifications

About this item

RS Pro, stainless steel threaded fittings, are suitable for applications requiring a secure and reliable pipe connection. Heavily used within the plumbing industry, water and gas will flow through in a tightly sealed construction. These fittings are made from a durable 316 stainless steel and incorporate a hexagon nipple to ensure a simple and secure connection for pipe and hose work. All models are highly reliable and excellent quality.

  • Hexagon nipple fitting
  • 316 Stainless steel
  • 150 lbs. fitting
  • With BSPT tapered screw end
  • Hexagon nipple is made from S/40 pipe

Applications: All throughout the home (bathroom, kitchen etc.) Any building with running water, heating or gas; Sewage and underground networks; Ships and boats; Car engines.