DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) ≥99% (by HPCE), solid
Catalog # D0815-100MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance (Colour):Red
- Appearance (Form):Powder
- Proton NMR spectrum:Conforms to Structure
- Solubility (Colour):Orange
- Solubility (Turbidity)
0.5 mg/ml, DMSO:Clear - EmM
in MeOH:36.0 - 41.0 - Wavelength:457 - 463 nm
- Purity (HPLC):≥99%
Specification Test Results
About this item
:
:380.273 g/mol
CAS Number:3785-01-1
Formula:C₁₇H₂₁IN₂
MDL Number:MFCD00011994