Specifications
- Description:Label printer, colour, BradyJet J4000-UK-BWSSFID, with safety and facility ID software, UK-plug
- Model:J4000-UK-BWSSFID
- Print resolution:4800 dpi
- Materials supported:Continuous and die-cut
- Computer requirements:PC
- Print width:209,55 mm (max.)
- Power supply:AC adaptor
- Connections:USB; ethernet
- Printer:Full Colour
- Print method:Inkjet
- Print speed:5000 max. labels/day
- Pk:1
- For:Facility identification; general identification; hazard communication identification; lean and 5S labelling; pipe and valve marking; warehouse marking
Specifications
About this item
This industry inkjet workhorse creates up to 5000 impactful full-colour labels per day up to 203,20 mm wide on continuous or die-cut material.
- Full colour
- Long-lasting
- 5000 labels per day
Ideal for a wide variety of applications in safety, including lean/5S, lockout tagout, instructional or safety chemical labelling for handling, storage or transport, as well as lab settings. Contains the colour options you need to create ANSI, GHS, ISO, BS5609 and arc flash compliant labels. Streamline storage by only needing to stock white label material and print only what you need, when you need it, in photo-quality full colour every time.
Easily print PDFs or from third-party software packages with the optimised windows driver. Includes Brady workstation safety and facility identification software suite, enabling you to immediately start designing professional signs and labels.