VWR® Disposable Cleanroom Socks
114-9707
- Clothing Size:One size
- Garment application:Cleanroom,Laboratory,Clinical
- Colour:White
- Description:VWR® Disposable cleanroom socks
- Pk:400 PAIR
- Fabric:Poly/Nylon blend
VWR® Disposable cleanroom socks are designed to provide all day comfort inside a cleanroom. They are made from materials that prevent any particle generation while in use.
- Single-use disposable socks
- Poly/Nylon blend
- Comfortable for extended use
- Cleanroom compatible
- Annex 1 compliant
Cleanroom socks provide comfort and durability for limited and extended wear in cleanroom environments. Suitable for use in ISO 4 to 6 (Class 10 to 1000) cleanrooms. Also available in a launderable format (see part numbers VWRI113-9936 - VWRI113-9939).
Used disposable socks can be recycled using the Fabrics and Clothing Zero Waste Box™ from TerraCycle.
: 400 pairs per case.